missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diacetoxytoluene 97.0+%, TCI America™

Catalog No. D331825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D331825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D331825G Supplier TCI America Supplier No. D331825G
Only null left

Chemical Identifiers

CAS 717-27-1
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
InChI Key KUZVIVNLNXNLAQ-UHFFFAOYSA-N
Synonym 2,5-diacetoxytoluene, methylhydroquinone diacetate, 1,4-diacetoxy-2-methylbenzene, 1,4-benzenediol,2-methyl-, 1,4-diacetate, 4-acetyloxy-3-methylphenyl acetate, acmc-209ojx, methylhydroquinone, diacetate, 2-methylhydroquinone diacetate, paragos 310054
PubChem CID 4184462
IUPAC Name (4-acetyloxy-3-methylphenyl) acetate
SMILES CC1=C(C=CC(=C1)OC(=O)C)OC(=O)C

Specifications

Melting Point 49°C
Color White-Yellow
Quantity 25 g
Formula Weight 208.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Diacetoxytoluene