accessibility menu, dialog, popup

UserName

CAS RN 717-27-1

CH3OOOCH3OH3C

CAS RN 717-27-1

IUPAC Name: 4-(acetyloxy)-2-methylphenyl acetate
Synonyms: 4-(acetyloxy)-2-methylphenyl acetate
Molecular Weight (g/mol): 208.21
Molecular Formula: C11H12O4
InChi Key: KUZVIVNLNXNLAQ-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C(OC(C)=O)C(C)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results