missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(4-bromophenyl)-1,1-dimethyl-3,4-diphenylsilole 95.0+%, TCI America™

Catalog No. B45471G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45471G 1 g
B45475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45471G Supplier TCI America Supplier No. B45471G

Chemical Identifiers

CAS 866769-99-5
Molecular Formula C30H24Br2Si
Molecular Weight (g/mol) 572.42
MDL Number MFCD28384134
InChI Key ZIHPPFVYCIHQLQ-UHFFFAOYSA-N
Synonym 2,5-Bis(4-bromophenyl)-1,1-dimethyl-3,4-diphenylsilacyclopenta-2,4-diene
PubChem CID 91972159
IUPAC Name 2,5-bis(4-bromophenyl)-1,1-dimethyl-3,4-diphenyl-1H-silole
SMILES C[Si]1(C)C(=C(C(=C1C1=CC=C(Br)C=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(Br)C=C1

Specifications

Melting Point 224°C
Color Yellow
Quantity 1 g
Formula Weight 572.42
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Bis(4-bromophenyl)-1,1-dimethyl-3,4-diphenylsilole