CAS RN 866769-99-5
IUPAC Name:
2,5-bis(4-bromophenyl)-1,1-dimethyl-3,4-diphenyl-1H-silole
Synonyms:
2,5-bis(4-bromophenyl)-1,1-dimethyl-3,4-diphenylsilole
Molecular Weight (g/mol):
572.42
Molecular Formula:
C30H24Br2Si
InChi Key:
ZIHPPFVYCIHQLQ-UHFFFAOYSA-N
SMILES:
C[Si]1(C)C(=C(C(=C1C1=CC=C(Br)C=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(Br)C=C1
1
–
1
of
1
results
1
2,5-Bis(4-bromophenyl)-1,1-dimethyl-3,4-diphenylsilole 95.0+%, TCI America™
CAS: 866769-99-5 Molecular Formula: C30H24Br2Si Molecular Weight (g/mol): 572.42 MDL Number: MFCD28384134 InChI Key: ZIHPPFVYCIHQLQ-UHFFFAOYSA-N Synonym: 2,5-Bis(4-bromophenyl)-1,1-dimethyl-3,4-diphenylsilacyclopenta-2,4-diene PubChem CID: 91972159 IUPAC Name: 2,5-bis(4-bromophenyl)-1,1-dimethyl-3,4-diphenyl-1H-silole SMILES: C[Si]1(C)C(=C(C(=C1C1=CC=C(Br)C=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(Br)C=C1