missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-tert-Butylphenyl)pyridine 97.0+%, TCI America™
$290.79 - $1,002.86
Chemical Identifiers
| CAS | 524713-66-4 |
|---|---|
| Molecular Formula | C15H17N |
| Molecular Weight (g/mol) | 211.31 |
| MDL Number | MFCD07775634 |
| InChI Key | RHXQSMVSBYAGQV-UHFFFAOYSA-N |
| PubChem CID | 12984537 |
| IUPAC Name | 2-(4-tert-butylphenyl)pyridine |
| SMILES | CC(C)(C)C1=CC=C(C=C1)C1=CC=CC=N1 |
Chemical Identifiers
| 524713-66-4 | |
| 211.31 | |
| RHXQSMVSBYAGQV-UHFFFAOYSA-N | |
| 2-(4-tert-butylphenyl)pyridine |
| C15H17N | |
| MFCD07775634 | |
| 12984537 | |
| CC(C)(C)C1=CC=C(C=C1)C1=CC=CC=N1 |
Specifications
| 524713-66-4 | |
| C15H17N | |
| 200 mg | |
| CC(C)(C)C1=CC=C(C=C1)C1=CC=CC=N1 | |
| 211.31 | |
| 211.31 | |
| Liquid |
| Yellow | |
| MFCD07775634 | |
| RHXQSMVSBYAGQV-UHFFFAOYSA-N | |
| 2-(4-tert-butylphenyl)pyridine | |
| 12984537 | |
| ≥97.0% (GC) | |
| 2-(4-tert-Butylphenyl)pyridine |
Safety and Handling
TSCA : No