accessibility menu, dialog, popup

UserName

CH3H3CCH3N

CAS RN 524713-66-4

IUPAC Name: 2-(4-tert-butylphenyl)pyridine
Synonyms: 2-(4-tert-butylphenyl)pyridine
Molecular Weight (g/mol): 211.31
Molecular Formula: C15H17N
InChi Key: RHXQSMVSBYAGQV-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C1=CC=CC=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results