missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-tert-Butylphenyl)pyridine 97.0+%, TCI America™
Supplier: TCI America B40921G
Specifications
| 2-(4-tert-Butylphenyl)pyridine | |
| Yellow | |
| MFCD07775634 | |
| RHXQSMVSBYAGQV-UHFFFAOYSA-N | |
| 2-(4-tert-butylphenyl)pyridine | |
| 12984537 | |
| ≥97.0% (GC) |
| 524713-66-4 | |
| C15H17N | |
| 1 g | |
| CC(C)(C)C1=CC=C(C=C1)C1=CC=CC=N1 | |
| 211.31 | |
| 211.31 | |
| Liquid |
Chemical Identifiers
| 524713-66-4 | |
| 211.31 | |
| RHXQSMVSBYAGQV-UHFFFAOYSA-N | |
| 2-(4-tert-butylphenyl)pyridine |
| C15H17N | |
| MFCD07775634 | |
| 12984537 | |
| CC(C)(C)C1=CC=C(C=C1)C1=CC=CC=N1 |
Safety and Handling
TSCA : No