missing translation for 'onlineSavingsMsg'
Learn More

2-(4-tert-Butylphenyl)ethanol, 96%

Catalog No. AAL1361106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1361106 5 g
AAL1361114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1361106 Supplier Thermo Scientific Chemicals Supplier No. L1361106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5406-86-0 | C12H18O | 178.275 g/mol

2-(4-tert-Butylphenyl)ethanol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-tert-Butylphenyl)ethanol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 5406-86-0
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.275
MDL Number MFCD00236023
InChI Key NZGMMENPUKHODD-UHFFFAOYSA-N
Synonym 2-4-tert-butylphenyl ethanol, 2-4-tert-butyl phenyl ethanol, p-tert-butylphenethyl alcohol, p-tert-butyl phenethyl alcohol, phenethyl alcohol, p-tert-butyl, 4-tert-butylphenethyl alcohol, unii-aes74i7nru, aes74i7nru, benzeneethanol, 4-1,1-dimethylethyl
PubChem CID 79410
IUPAC Name 2-(4-tert-butylphenyl)ethanol
SMILES CC(C)(C)C1=CC=C(C=C1)CCO

Specifications

Melting Point 31°C to 35°C
Density 0.975
Boiling Point 141°C to 143°C (15 mmHg)
Refractive Index 1.5209
Quantity 5 g
UN Number UN3077
Beilstein 2246595
Solubility Information Insoluble in water.
Formula Weight 178.28
Percent Purity 96%
Chemical Name or Material 2-(4-tert-Butylphenyl)ethanol
Show More Show Less
Hazard Category H318-H361f-H373
Hazard Statement GHS H Statement
H318-H361-H373
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P260-P281-P305+P351+P338-P308+P313-P310-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 410-020-5
RTECSNumber SG7875000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only