accessibility menu, dialog, popup

UserName

CAS RN 5406-86-0

IUPAC Name: 2-(4-tert-butylphenyl)ethan-1-ol
Synonyms: 2-(4-tert-butylphenyl)ethanol
Molecular Weight (g/mol): 178.28
Molecular Formula: C12H18O
InChi Key: NZGMMENPUKHODD-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(CCO)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results