missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Nitrophenyl)ethyl Bromide 98.0+%, TCI America™
$156.59 - $488.38
Chemical Identifiers
| CAS | 5339-26-4 |
|---|---|
| Molecular Formula | C8H8BrNO2 |
| Molecular Weight (g/mol) | 230.061 |
| MDL Number | MFCD00007386 |
| InChI Key | NTURQZFFJDCTMZ-UHFFFAOYSA-N |
| Synonym | 4-nitrophenethyl bromide, 1-2-bromoethyl-4-nitrobenzene, benzene, 1-2-bromoethyl-4-nitro, 4-nitrophenethylbromide, p-nitrophenethyl bromide, 2-4-nitrophenyl ethyl bromide, 4-2-bromoethyl nitrobenzene, 4-nitrophenylethyl bromide, .beta.-p-nitrophenyl ethyl bromide, phenethylbromide4nitro |
| PubChem CID | 79266 |
| IUPAC Name | 1-(2-bromoethyl)-4-nitrobenzene |
| SMILES | C1=CC(=CC=C1CCBr)[N+](=O)[O-] |
Chemical Identifiers
| 5339-26-4 | |
| 230.061 | |
| NTURQZFFJDCTMZ-UHFFFAOYSA-N | |
| 79266 | |
| C1=CC(=CC=C1CCBr)[N+](=O)[O-] |
| C8H8BrNO2 | |
| MFCD00007386 | |
| 4-nitrophenethyl bromide, 1-2-bromoethyl-4-nitrobenzene, benzene, 1-2-bromoethyl-4-nitro, 4-nitrophenethylbromide, p-nitrophenethyl bromide, 2-4-nitrophenyl ethyl bromide, 4-2-bromoethyl nitrobenzene, 4-nitrophenylethyl bromide, .beta.-p-nitrophenyl ethyl bromide, phenethylbromide4nitro | |
| 1-(2-bromoethyl)-4-nitrobenzene |
Specifications
| 5339-26-4 | |
| C8H8BrNO2 | |
| 25 g | |
| NTURQZFFJDCTMZ-UHFFFAOYSA-N | |
| 1-(2-bromoethyl)-4-nitrobenzene | |
| 79266 | |
| ≥98.0% (GC) | |
| 2-(4-Nitrophenyl)ethyl Bromide |
| 69°C | |
| MFCD00007386 | |
| 4-nitrophenethyl bromide, 1-2-bromoethyl-4-nitrobenzene, benzene, 1-2-bromoethyl-4-nitro, 4-nitrophenethylbromide, p-nitrophenethyl bromide, 2-4-nitrophenyl ethyl bromide, 4-2-bromoethyl nitrobenzene, 4-nitrophenylethyl bromide, .beta.-p-nitrophenyl ethyl bromide, phenethylbromide4nitro | |
| C1=CC(=CC=C1CCBr)[N+](=O)[O-] | |
| 230.061 | |
| 230.06 | |
| Crystalline Powder |
Safety and Handling
TSCA : No