accessibility menu, dialog, popup

UserName

ONOBr

CAS RN 5339-26-4

IUPAC Name: 1-(2-bromoethyl)-4-nitrobenzene
Synonyms: PHENETHYLBROMIDE4nitro
Molecular Weight (g/mol): 230.06
Molecular Formula: C8H8BrNO2
InChi Key: NTURQZFFJDCTMZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(CCBr)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)