missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Nitrophenyl)ethyl Bromide 98.0+%, TCI America™

Catalog No. N059125G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N059125G 25 g
N0591100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N059125G Supplier TCI America Supplier No. N059125G

Chemical Identifiers

CAS 5339-26-4
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.061
MDL Number MFCD00007386
InChI Key NTURQZFFJDCTMZ-UHFFFAOYSA-N
Synonym 4-nitrophenethyl bromide, 1-2-bromoethyl-4-nitrobenzene, benzene, 1-2-bromoethyl-4-nitro, 4-nitrophenethylbromide, p-nitrophenethyl bromide, 2-4-nitrophenyl ethyl bromide, 4-2-bromoethyl nitrobenzene, 4-nitrophenylethyl bromide, .beta.-p-nitrophenyl ethyl bromide, phenethylbromide4nitro
PubChem CID 79266
IUPAC Name 1-(2-bromoethyl)-4-nitrobenzene
SMILES C1=CC(=CC=C1CCBr)[N+](=O)[O-]

Specifications

Melting Point 69°C
Quantity 25 g
Formula Weight 230.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Nitrophenyl)ethyl Bromide