missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Methoxyphenyl)-4,6-bis(trichloromethyl)-1,3,5-triazine 98.0+%, TCI America™
Supplier: TCI America M124525G
Specifications
| 2-(4-Methoxyphenyl)-4,6-bis(trichloromethyl)-1,3,5-triazine | |
| 145°C | |
| C12H7Cl6N3O | |
| 25 g | |
| QRHHZFRCJDAUNA-UHFFFAOYSA-N | |
| 2-(4-methoxyphenyl)-4,6-bis(trichloromethyl)-1,3,5-triazine | |
| 19163 | |
| ≥98.0% (GC) |
| 3584-23-4 | |
| Yellow | |
| MFCD00521486 | |
| 4,6-Bis(trichloromethyl)-2-(4-methoxyphenyl)-1,3,5-triazine | |
| COC1=CC=C(C=C1)C2=NC(=NC(=N2)C(Cl)(Cl)Cl)C(Cl)(Cl)Cl | |
| 421.908 | |
| 421.91 | |
| Crystalline Powder |
Chemical Identifiers
| 3584-23-4 | |
| 421.908 | |
| QRHHZFRCJDAUNA-UHFFFAOYSA-N | |
| 19163 | |
| COC1=CC=C(C=C1)C2=NC(=NC(=N2)C(Cl)(Cl)Cl)C(Cl)(Cl)Cl |
| C12H7Cl6N3O | |
| MFCD00521486 | |
| 4,6-Bis(trichloromethyl)-2-(4-methoxyphenyl)-1,3,5-triazine | |
| 2-(4-methoxyphenyl)-4,6-bis(trichloromethyl)-1,3,5-triazine |
Safety and Handling
TSCA : No