missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methoxyphenyl)-4,6-bis(trichloromethyl)-1,3,5-triazine 98.0+%, TCI America™

Catalog No. p-7132614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12455G 5 g
M124525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12455G Supplier TCI America Supplier No. M12455G

Chemical Identifiers

CAS 3584-23-4
Molecular Formula C12H7Cl6N3O
Molecular Weight (g/mol) 421.908
MDL Number MFCD00521486
InChI Key QRHHZFRCJDAUNA-UHFFFAOYSA-N
Synonym 4,6-Bis(trichloromethyl)-2-(4-methoxyphenyl)-1,3,5-triazine
PubChem CID 19163
IUPAC Name 2-(4-methoxyphenyl)-4,6-bis(trichloromethyl)-1,3,5-triazine
SMILES COC1=CC=C(C=C1)C2=NC(=NC(=N2)C(Cl)(Cl)Cl)C(Cl)(Cl)Cl

Specifications

Melting Point 145°C
Color Yellow
Quantity 5 g
Formula Weight 421.91
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Methoxyphenyl)-4,6-bis(trichloromethyl)-1,3,5-triazine