missing translation for 'onlineSavingsMsg'
Learn More

2-[(4-Methoxybenzylidene)amino]fluorene 98.0+%, TCI America™

Catalog No. M05865G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M05865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M05865G Supplier TCI America Supplier No. M05865G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5424-78-2
Molecular Formula C21H17NO
Molecular Weight (g/mol) 299.373
MDL Number MFCD00019064
InChI Key AFAJHWLHSQIELD-UHFFFAOYSA-N
Synonym 2-(p-Anisalamino)fluorene, 2-[(p-Anisylidene)amino]fluorene
PubChem CID 224057
IUPAC Name N-(9H-fluoren-2-yl)-1-(4-methoxyphenyl)methanimine
SMILES COC1=CC=C(C=C1)C=NC2=CC3=C(C=C2)C4=CC=CC=C4C3

Specifications

Color Yellow
Quantity 5 g
Formula Weight 299.37
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-[(4-Methoxybenzylidene)amino]fluorene