CAS RN 5424-78-2
IUPAC Name:
N-(9H-fluoren-2-yl)-1-(4-methoxyphenyl)methanimine
Synonyms:
N-(9H-fluoren-2-yl)-1-(4-methoxyphenyl)methanimine
Molecular Weight (g/mol):
299.37
Molecular Formula:
C21H17NO
InChi Key:
AFAJHWLHSQIELD-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=NC2=CC=C3C(CC4=CC=CC=C34)=C2)C=C1
1
–
1
of
1
results
1
2-[(4-Methoxybenzylidene)amino]fluorene 98.0+%, TCI America™
CAS: 5424-78-2 Molecular Formula: C21H17NO Molecular Weight (g/mol): 299.373 MDL Number: MFCD00019064 InChI Key: AFAJHWLHSQIELD-UHFFFAOYSA-N Synonym: 2-(p-Anisalamino)fluorene, 2-[(p-Anisylidene)amino]fluorene PubChem CID: 224057 IUPAC Name: N-(9H-fluoren-2-yl)-1-(4-methoxyphenyl)methanimine SMILES: COC1=CC=C(C=C1)C=NC2=CC3=C(C=C2)C4=CC=CC=C4C3