missing translation for 'onlineSavingsMsg'
Learn More

2,4-Hexadiyne-1,6-diol 99.0+%, TCI America™

Supplier:  TCI America H06191G

 View more versions of this product

Catalog No. H06191G


Only null left

Description

Description

Specifications

Specifications

2,4-Hexadiyne-1,6-diol
113°C
C6H6O2
1 g
2,4-hexadiyne-1,6-diol, diacetylene glycol, 2,4-hexadiynediol, 2,4-hexadiyn-1,6-diol, 2,6-diol, acmc-1ahii, hoch2c=c-c=cch2oh, 1,6-dihydroxy-2,4-hexadiyne, hexa-2, 4-diyne-1, 6-diol, jxmqykbazrdvtc-uhfffaoysa
C(C#CC#CCO)O
110.112
110.11
Crystalline Powder
3031-68-3
White-Yellow
MFCD00020619
1325
JXMQYKBAZRDVTC-UHFFFAOYSA-N
hexa-2,4-diyne-1,6-diol
94973
≥99.0% (GC)

Chemical Identifiers

3031-68-3
110.112
JXMQYKBAZRDVTC-UHFFFAOYSA-N
94973
C(C#CC#CCO)O
C6H6O2
MFCD00020619
2,4-hexadiyne-1,6-diol, diacetylene glycol, 2,4-hexadiynediol, 2,4-hexadiyn-1,6-diol, 2,6-diol, acmc-1ahii, hoch2c=c-c=cch2oh, 1,6-dihydroxy-2,4-hexadiyne, hexa-2, 4-diyne-1, 6-diol, jxmqykbazrdvtc-uhfffaoysa
hexa-2,4-diyne-1,6-diol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions