accessibility menu, dialog, popup

UserName

OHHO

CAS RN 3031-68-3

IUPAC Name: hexa-2,4-diyne-1,6-diol
Synonyms: hexa-2,4-diyne-1,6-diol
Molecular Weight (g/mol): 110.11
Molecular Formula: C6H6O2
InChi Key: JXMQYKBAZRDVTC-UHFFFAOYSA-N
SMILES: OCC#CC#CCO
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)