missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Ethoxyphenyl)-2-methylpropanol 98.0+%, TCI America™
Supplier: TCI America E073025G
Specifications
| 2-(4-Ethoxyphenyl)-2-methylpropanol | |
| 48°C | |
| C12H18O2 | |
| 25 g | |
| CCOC1=CC=C(C=C1)C(C)(C)CO | |
| 194.27 | |
| CHEBI:39350 | |
| ≥98.0% (GC) |
| 83493-63-4 | |
| White-Yellow | |
| MFCD07787279 | |
| OZEZBKUHAGFQME-UHFFFAOYSA-N | |
| 2-(4-ethoxyphenyl)-2-methylpropan-1-ol | |
| 158525 | |
| 194.27 | |
| Crystalline Powder |
Chemical Identifiers
| 83493-63-4 | |
| 194.27 | |
| OZEZBKUHAGFQME-UHFFFAOYSA-N | |
| CHEBI:39350 | |
| CCOC1=CC=C(C=C1)C(C)(C)CO |
| C12H18O2 | |
| MFCD07787279 | |
| 158525 | |
| 2-(4-ethoxyphenyl)-2-methylpropan-1-ol |
Safety and Handling
TSCA : No