missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Ethoxyphenyl)-2-methylpropanol 98.0+%, TCI America™

Catalog No. p-7129119
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07305G 5 g
E073025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07305G Supplier TCI America Supplier No. E07305G

Chemical Identifiers

CAS 83493-63-4
Molecular Formula C12H18O2
Molecular Weight (g/mol) 194.27
MDL Number MFCD07787279
InChI Key OZEZBKUHAGFQME-UHFFFAOYSA-N
PubChem CID 158525
ChEBI CHEBI:39350
IUPAC Name 2-(4-ethoxyphenyl)-2-methylpropan-1-ol
SMILES CCOC1=CC=C(C=C1)C(C)(C)CO

Specifications

Melting Point 48°C
Color White-Yellow
Quantity 5 g
Formula Weight 194.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Ethoxyphenyl)-2-methylpropanol