missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrophenol (wetted with ca. 20% Water) (unit weight on dry weight basis) 98.0+%, TCI America™

Catalog No. D010925G
Change view
Click to view available options
Quantity:
25 g
300 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D010925G 25 g
D0109300G 300 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D010925G Supplier TCI America Supplier No. D010925G

Chemical Identifiers

CAS 51-28-5
Molecular Formula C6H4N2O5
Molecular Weight (g/mol) 184.107
MDL Number MFCD00007115
InChI Key UFBJCMHMOXMLKC-UHFFFAOYSA-N
Synonym 2,4-dinitrophenol, aldifen, solfo black b, 2,4-dnp, tertrosulphur pbr, phenol, 2,4-dinitro, alpha-dinitrophenol, solfo black g, 1-hydroxy-2,4-dinitrobenzene, nitro kleenup
PubChem CID 1493
ChEBI CHEBI:42017
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])O

Specifications

Melting Point 113°C
Quantity 25 g
UN Number 1320
Formula Weight 184.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dinitrophenol (wetted with ca. 20% Water) (unit weight on dry weight basis)
EINECSNumber (3)-0797
RTECSNumber SL2800000
TSCA Yes