missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrofluorobenzene 99.0+%, TCI America™

Catalog No. D083525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D083525G 25 g
D0835500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D083525G Supplier TCI America Supplier No. D083525G

Chemical Identifiers

CAS 70-34-8
Molecular Formula C6H3FN2O4
Molecular Weight (g/mol) 186.098
MDL Number MFCD00007056
InChI Key LOTKRQAVGJMPNV-UHFFFAOYSA-N
Synonym 2,4-dinitrofluorobenzene, dinitrofluorobenzene, fdnb, dnfb, 2,4-dinitro-1-fluorobenzene, sanger's reagent, fluoro-2,4-dinitrobenzene, 2,4-dinitrobenzenefluoride, 2,4-dinitrophenyl fluoride, fluorodinitrobenzene
PubChem CID 6264
ChEBI CHEBI:53049
IUPAC Name 1-fluoro-2,4-dinitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])F

Specifications

Boiling Point 296°C
Quantity 25 g
UN Number 2811
Formula Weight 186.10
Percent Purity ≥99.0% (GC)
Physical Form Crystals
Chemical Name or Material 2,4-Dinitrofluorobenzene
EINECSNumber (3)-0456
RTECSNumber CZ7800000
TSCA Yes