Learn More
2,4-Dinitrobenzaldehyde, 98%
CAS: 528-75-6 | C7H4N2O5 | 196.118 g/mol
Supplier: Thermo Scientific Chemicals A1545206
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2,4-Dinitrobenzaldehyde | |
| 65°C to 68°C | |
| C7H4N2O5 | |
| 5 g | |
| Air Sensitive | |
| benzaldehyde, 2,4-dinitro, unii-zac919yu3z, ccris 2684, zac919yu3z, pubchem8223, benzaldehyde,4-dinitro, 2,4-dinitro-benzaldehyde, acmc-209l2f, 3-07-00-00923 beilstein handbook reference, ksc490k9l | |
| C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C=O | |
| 196.118 | |
| 196.12 |
| 528-75-6 | |
| 190°C (10 mmHg) | |
| MFCD00013376 | |
| 1878706 | |
| 14,3272 | |
| ZILXIZUBLXVYPI-UHFFFAOYSA-N | |
| 2,4-dinitrobenzaldehyde | |
| 68250 | |
| 98% |
Chemical Identifiers
| 528-75-6 | |
| 196.118 | |
| ZILXIZUBLXVYPI-UHFFFAOYSA-N | |
| 68250 | |
| C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C=O |
| C7H4N2O5 | |
| MFCD00013376 | |
| benzaldehyde, 2,4-dinitro, unii-zac919yu3z, ccris 2684, zac919yu3z, pubchem8223, benzaldehyde,4-dinitro, 2,4-dinitro-benzaldehyde, acmc-209l2f, 3-07-00-00923 beilstein handbook reference, ksc490k9l | |
| 2,4-dinitrobenzaldehyde |
Safety and Handling
GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 208-440-7
RTECSNumber : CU5957000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only