missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrobenzaldehyde, 98%

Catalog No. p-7022598
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1545203 1 g
AAA1545206 5 g
AAA1545214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1545203 Supplier Thermo Scientific Chemicals Supplier No. A1545203

CAS: 528-75-6 | C7H4N2O5 | 196.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 528-75-6
Molecular Formula C7H4N2O5
Molecular Weight (g/mol) 196.118
MDL Number MFCD00013376
InChI Key ZILXIZUBLXVYPI-UHFFFAOYSA-N
Synonym benzaldehyde, 2,4-dinitro, unii-zac919yu3z, ccris 2684, zac919yu3z, pubchem8223, benzaldehyde,4-dinitro, 2,4-dinitro-benzaldehyde, acmc-209l2f, 3-07-00-00923 beilstein handbook reference, ksc490k9l
PubChem CID 68250
IUPAC Name 2,4-dinitrobenzaldehyde
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C=O

Specifications

Melting Point 65°C to 68°C
Boiling Point 190°C (10 mmHg)
Quantity 1 g
Beilstein 1878706
Sensitivity Air Sensitive
Merck Index 14,3272
Formula Weight 196.12
Percent Purity 98%
Chemical Name or Material 2,4-Dinitrobenzaldehyde
Hazard Category Warning
Hazard Statement GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 208-440-7
RTECSNumber CU5957000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only