missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylthiophenol, 95%

CAS: 13616-82-5 | C8H10S | 138.228 g/mol

Supplier:  Thermo Scientific Chemicals B2316114

 View more versions of this product

Catalog No. AAB2316114

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,4-Dimethylthiophenol
1.02
87°C (189°F)
C8H10S
MFCD00010019
UN2810
2,4-dimethylthiophenol, 2,4-dimethyl thiophenol, benzenethiol, 2,4-dimethyl, 2,4-thioxylenol, 2,4-xylenethiol, 4-mercapto-m-xylene, 2,4-dimethylbenzene-1-thiol, 4-methylthiocresol, 4-thio-m-xylene, m-xylene-4-thiol
CC1=CC(=C(C=C1)S)C
138.228
138.23
13616-82-5
207°C to 208°C
Unpleasant
1.569
25 g
Air Sensitive
AMNLXDDJGGTIPL-UHFFFAOYSA-N
2,4-dimethylbenzenethiol
83617
95%

Chemical Identifiers

13616-82-5
138.228
AMNLXDDJGGTIPL-UHFFFAOYSA-N
83617
CC1=CC(=C(C=C1)S)C
C8H10S
MFCD00010019
2,4-dimethylthiophenol, 2,4-dimethyl thiophenol, benzenethiol, 2,4-dimethyl, 2,4-thioxylenol, 2,4-xylenethiol, 4-mercapto-m-xylene, 2,4-dimethylbenzene-1-thiol, 4-methylthiocresol, 4-thio-m-xylene, m-xylene-4-thiol
2,4-dimethylbenzenethiol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Hazard Class: 3; Packaging Group: III

EINECSNumber : 237-100-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only