accessibility menu, dialog, popup

UserName

CAS RN 13616-82-5

IUPAC Name: 2,4-dimethylbenzene-1-thiol
Synonyms: 2,4-dimethylbenzenethiol
Molecular Weight (g/mol): 138.23
Molecular Formula: C8H10S
InChi Key: AMNLXDDJGGTIPL-UHFFFAOYSA-N
SMILES: CC1=CC=C(S)C(C)=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)