missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylphenol 98.0+%, TCI America™

Catalog No. D077425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D077425G 25 g
D0774500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D077425G Supplier TCI America Supplier No. D077425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 105-67-9
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00002233
InChI Key KUFFULVDNCHOFZ-UHFFFAOYSA-N
Synonym 2,4-xylenol, phenol, 2,4-dimethyl, gallex, 4,6-dimethylphenol, 1-hydroxy-2,4-dimethylbenzene, 4-hydroxy-1,3-dimethylbenzene, rcra waste number u101, 2,4-dimethyl phenol, caswell no. 907a, lysol brand disinfectant
PubChem CID 7771
ChEBI CHEBI:34241
IUPAC Name 2,4-dimethylphenol
SMILES CC1=CC(=C(C=C1)O)C

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
UN Number 3430
Boiling Point 212°C
Formula Weight 122.17
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,4-Dimethylphenol
EINECSNumber (3)-0521&(4)-0057
RTECSNumber ZE5600000
TSCA Yes