accessibility menu, dialog, popup

UserName

CH3OHH3C

CAS RN 105-67-9

IUPAC Name: 2,4-dimethylphenol
Synonyms: gallex lysol brand disinfectant bulk lysol brand disinfectant caswell no. 907A ccris 721 rcra waste number U101 bacticin epa pesticide chemical code 086804 du cor concentrated fly insecticide
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: KUFFULVDNCHOFZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C(C)=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)