missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylphenol, 99%

CAS: 105-67-9 | C8H10O | 122.17 g/mol

Supplier:  Thermo Scientific Chemicals 408451000

 View more versions of this product

Catalog No. AC408451000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 4-Dimethylphenol
105-67-9
98.5
1.0270g/mL
100 g
Glass bottle
1.5375 to 1.5405
1.027
(CH3)2C6H3OH
06, 486
2,4-xylenol, phenol, 2,4-dimethyl, gallex, 4,6-dimethylphenol, 1-hydroxy-2,4-dimethylbenzene, 4-hydroxy-1,3-dimethylbenzene, rcra waste number u101, 2,4-dimethyl phenol, caswell no. 907a, lysol brand disinfectant
KUFFULVDNCHOFZ-UHFFFAOYSA-N
2,4-dimethylphenol
7771
122.17
21.0°C to 23.0°C
Colorless to Yellow
100.0
211.0°C
Authentic
Liquid
98.5% min. (GC)
C8H10O
MFCD00002233
15, 10279
Solubility in water: 0.5% (25°C). Other solubilities: freely soluble in alcohol,chloroform,ether,benz,ene,soluble in naoh solutions
CC1=CC(=C(C=C1)O)C
122.17
CHEBI:34241
99%

Chemical Identifiers

105-67-9
122.17
KUFFULVDNCHOFZ-UHFFFAOYSA-N
7771
2,4-dimethylphenol
C8H10O
MFCD00002233
2,4-xylenol, phenol, 2,4-dimethyl, gallex, 4,6-dimethylphenol, 1-hydroxy-2,4-dimethylbenzene, 4-hydroxy-1,3-dimethylbenzene, rcra waste number u101, 2,4-dimethyl phenol, caswell no. 907a, lysol brand disinfectant
CHEBI:34241
CC1=CC(=C(C=C1)O)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit

GHS Signal Word: Danger

EINECSNumber : 203-321-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only