Learn More
2,4-Dimethylphenol, 99%
CAS: 105-67-9 | C8H10O | 122.17 g/mol
Supplier: Thermo Scientific Chemicals 408450250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2, 4-Dimethylphenol | |
| 105-67-9 | |
| 98.5 | |
| 1.0270g/mL | |
| 25 g | |
| Glass bottle | |
| 1.5375 to 1.5405 | |
| 1.027 | |
| (CH3)2C6H3OH | |
| 06, 486 | |
| 2,4-xylenol, phenol, 2,4-dimethyl, gallex, 4,6-dimethylphenol, 1-hydroxy-2,4-dimethylbenzene, 4-hydroxy-1,3-dimethylbenzene, rcra waste number u101, 2,4-dimethyl phenol, caswell no. 907a, lysol brand disinfectant | |
| KUFFULVDNCHOFZ-UHFFFAOYSA-N | |
| 2,4-dimethylphenol | |
| 7771 | |
| 122.17 |
| 21.0°C to 23.0°C | |
| Colorless to Yellow | |
| 100.0 | |
| 211.0°C | |
| Authentic | |
| Liquid | |
| 98.5% min. (GC) | |
| C8H10O | |
| MFCD00002233 | |
| 15, 10279 | |
| Solubility in water: 0.5% (25°C). Other solubilities: freely soluble in alcohol,chloroform,ether,benz,ene,soluble in naoh solutions | |
| CC1=CC(=C(C=C1)O)C | |
| 122.17 | |
| CHEBI:34241 | |
| 99% |
Chemical Identifiers
| 105-67-9 | |
| 122.17 | |
| KUFFULVDNCHOFZ-UHFFFAOYSA-N | |
| 7771 | |
| 2,4-dimethylphenol |
| C8H10O | |
| MFCD00002233 | |
| 2,4-xylenol, phenol, 2,4-dimethyl, gallex, 4,6-dimethylphenol, 1-hydroxy-2,4-dimethylbenzene, 4-hydroxy-1,3-dimethylbenzene, rcra waste number u101, 2,4-dimethyl phenol, caswell no. 907a, lysol brand disinfectant | |
| CHEBI:34241 | |
| CC1=CC(=C(C=C1)O)C |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit
GHS Signal Word: Danger
EINECSNumber : 203-321-6
RUO – Research Use Only