missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethylbenzonitrile 98.0+%, TCI America™
$124.89 - $374.68
Chemical Identifiers
| CAS | 21789-36-6 |
|---|---|
| Molecular Formula | C9H9N |
| Molecular Weight (g/mol) | 131.178 |
| MDL Number | MFCD00016337 |
| InChI Key | QLZDTHTXOUOSCV-UHFFFAOYSA-N |
| Synonym | 4-Cyano-m-xylene |
| PubChem CID | 89046 |
| IUPAC Name | 2,4-dimethylbenzonitrile |
| SMILES | CC1=CC(=C(C=C1)C#N)C |
Chemical Identifiers
| 21789-36-6 | |
| 131.178 | |
| QLZDTHTXOUOSCV-UHFFFAOYSA-N | |
| 89046 | |
| CC1=CC(=C(C=C1)C#N)C |
| C9H9N | |
| MFCD00016337 | |
| 4-Cyano-m-xylene | |
| 2,4-dimethylbenzonitrile |
Specifications
| 21789-36-6 | |
| 109°C | |
| Crystal-Lump at 20°C | |
| MFCD00016337 | |
| QLZDTHTXOUOSCV-UHFFFAOYSA-N | |
| 2,4-dimethylbenzonitrile | |
| 89046 | |
| ≥98.0% (GC) |
| White-Yellow | |
| 5 g | |
| C9H9N | |
| 4-Cyano-m-xylene | |
| CC1=CC(=C(C=C1)C#N)C | |
| 131.178 | |
| 131.18 | |
| 2,4-Dimethylbenzonitrile |
Safety and Handling
TSCA : No