missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethylbenzonitrile 98.0+%, TCI America™
Supplier: TCI America D449525G
Specifications
| 2,4-Dimethylbenzonitrile | |
| White-Yellow | |
| 25 g | |
| C9H9N | |
| 4-Cyano-m-xylene | |
| CC1=CC(=C(C=C1)C#N)C | |
| 131.178 | |
| 131.18 |
| 21789-36-6 | |
| 109°C | |
| Crystal-Lump at 20°C | |
| MFCD00016337 | |
| QLZDTHTXOUOSCV-UHFFFAOYSA-N | |
| 2,4-dimethylbenzonitrile | |
| 89046 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 21789-36-6 | |
| 131.178 | |
| QLZDTHTXOUOSCV-UHFFFAOYSA-N | |
| 89046 | |
| CC1=CC(=C(C=C1)C#N)C |
| C9H9N | |
| MFCD00016337 | |
| 4-Cyano-m-xylene | |
| 2,4-dimethylbenzonitrile |
Safety and Handling
TSCA : No