missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylbenzenethiol 96.0+%, TCI America™

Catalog No. p-7127424
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
D26485G 5 g
D264825G 25 g
2 options

Catalog No. D26485G

Supplier: TCI America D26485G

Only null left

Chemical Identifiers

CAS 13616-82-5
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD00010019
InChI Key AMNLXDDJGGTIPL-UHFFFAOYSA-N
Synonym 2,4-dimethylthiophenol, 2,4-dimethyl thiophenol, benzenethiol, 2,4-dimethyl, 2,4-thioxylenol, 2,4-xylenethiol, 4-mercapto-m-xylene, 2,4-dimethylbenzene-1-thiol, 4-methylthiocresol, 4-thio-m-xylene, m-xylene-4-thiol
PubChem CID 83617
IUPAC Name 2,4-dimethylbenzenethiol
SMILES CC1=CC(=C(C=C1)S)C

Specifications

Color Yellow
Boiling Point 214°C
Quantity 5 g
UN Number 1993
Formula Weight 138.23
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethylbenzenethiol
EINECSNumber (3)-1094
TSCA Yes