missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylaniline Hydrochloride 98.0+%, TCI America™

Catalog No. X002825G
Change view
Click to view available options
Quantity:
25 g
Catalog No. Quantity
X002825G 25 g
1 options

Catalog No. X002825G

Supplier: TCI America X002825G

Only null left

Chemical Identifiers

CAS 21436-96-4
Molecular Formula C8H12ClN
Molecular Weight (g/mol) 157.641
MDL Number MFCD00060209
InChI Key HFXISSJBRAPVLG-UHFFFAOYSA-N
Synonym m-Xylidine Hydrochloride
PubChem CID 88898
IUPAC Name 2,4-dimethylaniline;hydrochloride
SMILES CC1=CC(=C(C=C1)N)C.Cl

Specifications

Melting Point 238°C
Color White-Yellow
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 157.64
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material 2,4-Dimethylaniline Hydrochloride
EINECSNumber (3)-0129
RTECSNumber ZF0175000
TSCA No