accessibility menu, dialog, popup

UserName

HClCH3NH2H3C

CAS RN 21436-96-4

IUPAC Name: hydrogen 2,4-dimethylaniline chloride
Synonyms: 2,4-xylidine, hydrochloride
Molecular Weight (g/mol): 157.64
Molecular Formula: C8H12ClN
InChi Key: HFXISSJBRAPVLG-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC=C(N)C(C)=C1
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results