missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 33904-03-9 |
|---|---|
| Molecular Formula | C9H9NO2S |
| Molecular Weight (g/mol) | 195.24 |
| InChI Key | CNXSCEPTSXPBTP-UHFFFAOYSA-N |
| Synonym | 2,4-dimethoxyphenyl isothiocyanate, 1-isothiocyanato-2,4-dimethoxy-benzene, 2,4-dimethoxybenzenisothiocyanate, acmc-20aock, 2,4-dimethoxyphenylisothiocyanate, 1-isothiocyanato-2.4-dimethoxybenzene, isothiocyanic acid 2,4-dimethoxyphenyl ester |
| PubChem CID | 2736202 |
| IUPAC Name | 1-isothiocyanato-2,4-dimethoxybenzene |
| SMILES | COC1=CC(=C(C=C1)N=C=S)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC441780010
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 33904-03-9 | |
| 195.24 | |
| 2,4-dimethoxyphenyl isothiocyanate, 1-isothiocyanato-2,4-dimethoxy-benzene, 2,4-dimethoxybenzenisothiocyanate, acmc-20aock, 2,4-dimethoxyphenylisothiocyanate, 1-isothiocyanato-2.4-dimethoxybenzene, isothiocyanic acid 2,4-dimethoxyphenyl ester | |
| 1-isothiocyanato-2,4-dimethoxybenzene |
| C9H9NO2S | |
| CNXSCEPTSXPBTP-UHFFFAOYSA-N | |
| 2736202 | |
| COC1=CC(=C(C=C1)N=C=S)OC |
Specifications
| 33904-03-9 | |
| 100.0 | |
| 113°C | |
| 94% min. (GC) | |
| C9H9NO2S | |
| 2,4-dimethoxyphenyl isothiocyanate, 1-isothiocyanato-2,4-dimethoxy-benzene, 2,4-dimethoxybenzenisothiocyanate, acmc-20aock, 2,4-dimethoxyphenylisothiocyanate, 1-isothiocyanato-2.4-dimethoxybenzene, isothiocyanic acid 2,4-dimethoxyphenyl ester | |
| COC1=CC(=C(C=C1)N=C=S)OC | |
| 195.24 | |
| 195.24 | |
| 2, 4-Dimethoxyphenyl isothiocyanate |
| 94.0 | |
| 166.0°C to 167.0°C (6.0mmHg) | |
| Authentic | |
| Glass Bottle | |
| (CH3O)2C6H3NCS | |
| CNXSCEPTSXPBTP-UHFFFAOYSA-N | |
| 1-isothiocyanato-2,4-dimethoxybenzene | |
| 2736202 | |
| 95% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
RUO – Research Use Only