missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxybenzylamine, 98%

Catalog No. AAH2714006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2714006 5 g
AAH2714014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2714006 Supplier Thermo Scientific Chemicals Supplier No. H2714006

CAS: 20781-20-8 | C9H13NO2 | 167.208 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20781-20-8
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.208
MDL Number MFCD00052393
InChI Key QOWBXWFYRXSBAS-UHFFFAOYSA-N
PubChem CID 597250
IUPAC Name (2,4-dimethoxyphenyl)methanamine
SMILES COC1=CC(=C(C=C1)CN)OC

Specifications

Density 1.113
Boiling Point 140°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.549
Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Synonym 2,4-dimethoxybenzylamine, 2,4-dimethoxyphenyl methanamine, 2,4-dimethoxy benzylamine, 1-2,4-dimethoxyphenyl methanamine, benzenemethanamine, 2,4-dimethoxy, 2,4-dimethoxybenzyl amine, 2,4-dimethoxyphenyl methylamine, 2, 4-dimethoxybenzylamine, pubchem7398, 2,4dimethoxybenzylamine
Formula Weight 167.21
Percent Purity 98%
Chemical Name or Material 2,4-Dimethoxybenzylamine
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only