missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxybenzyl Alcohol 98.0+%, TCI America™

$48.46 - $229.29

Chemical Identifiers

CAS 7314-44-5
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00004614
InChI Key RNKOUSCCPHSCFE-UHFFFAOYSA-N
Synonym 2,4-dimethoxybenzyl alcohol, 2,4-dimethoxyphenyl methanol, benzenemethanol, 2,4-dimethoxy, 2,4-dimethoxybenzylalcohol, 1-hydroxymethyl-2,4-dimethoxybenzene, 2,4-dimethoxyphenyl methan-1-ol, pubchem7454, acmc-209opp, 2,4-dimethoxy-benzylalcohol, 2,4-dimethoxybenzyl-alcohol
PubChem CID 81750
IUPAC Name (2,4-dimethoxyphenyl)methanol
SMILES COC1=CC(=C(C=C1)CO)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D18355G
View Documents
TCI America
D18355G
5 g N/A
Only null left
 
D183525G
View Documents
TCI America
D183525G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7314-44-5
168.192
RNKOUSCCPHSCFE-UHFFFAOYSA-N
81750
COC1=CC(=C(C=C1)CO)OC
C9H12O3
MFCD00004614
2,4-dimethoxybenzyl alcohol, 2,4-dimethoxyphenyl methanol, benzenemethanol, 2,4-dimethoxy, 2,4-dimethoxybenzylalcohol, 1-hydroxymethyl-2,4-dimethoxybenzene, 2,4-dimethoxyphenyl methan-1-ol, pubchem7454, acmc-209opp, 2,4-dimethoxy-benzylalcohol, 2,4-dimethoxybenzyl-alcohol
(2,4-dimethoxyphenyl)methanol

Specifications

7314-44-5
White
C9H12O3
5 g
RNKOUSCCPHSCFE-UHFFFAOYSA-N
(2,4-dimethoxyphenyl)methanol
81750
≥98.0% (GC)
2,4-Dimethoxybenzyl Alcohol
36°C
135°C
MFCD00004614
2,4-dimethoxybenzyl alcohol, 2,4-dimethoxyphenyl methanol, benzenemethanol, 2,4-dimethoxy, 2,4-dimethoxybenzylalcohol, 1-hydroxymethyl-2,4-dimethoxybenzene, 2,4-dimethoxyphenyl methan-1-ol, pubchem7454, acmc-209opp, 2,4-dimethoxy-benzylalcohol, 2,4-dimethoxybenzyl-alcohol
COC1=CC(=C(C=C1)CO)OC
168.192
168.19
Crystals
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'