missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethoxybenzyl Alcohol 98.0+%, TCI America™
Supplier: TCI America D183525G
Specifications
| 2,4-Dimethoxybenzyl Alcohol | |
| 36°C | |
| 135°C | |
| MFCD00004614 | |
| 2,4-dimethoxybenzyl alcohol, 2,4-dimethoxyphenyl methanol, benzenemethanol, 2,4-dimethoxy, 2,4-dimethoxybenzylalcohol, 1-hydroxymethyl-2,4-dimethoxybenzene, 2,4-dimethoxyphenyl methan-1-ol, pubchem7454, acmc-209opp, 2,4-dimethoxy-benzylalcohol, 2,4-dimethoxybenzyl-alcohol | |
| COC1=CC(=C(C=C1)CO)OC | |
| 168.192 | |
| 168.19 | |
| Crystals |
| 7314-44-5 | |
| White | |
| C9H12O3 | |
| 25 g | |
| RNKOUSCCPHSCFE-UHFFFAOYSA-N | |
| (2,4-dimethoxyphenyl)methanol | |
| 81750 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 7314-44-5 | |
| 168.192 | |
| RNKOUSCCPHSCFE-UHFFFAOYSA-N | |
| 81750 | |
| COC1=CC(=C(C=C1)CO)OC |
| C9H12O3 | |
| MFCD00004614 | |
| 2,4-dimethoxybenzyl alcohol, 2,4-dimethoxyphenyl methanol, benzenemethanol, 2,4-dimethoxy, 2,4-dimethoxybenzylalcohol, 1-hydroxymethyl-2,4-dimethoxybenzene, 2,4-dimethoxyphenyl methan-1-ol, pubchem7454, acmc-209opp, 2,4-dimethoxy-benzylalcohol, 2,4-dimethoxybenzyl-alcohol | |
| (2,4-dimethoxyphenyl)methanol |
Safety and Handling
TSCA : No