missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxybenzenesulfonamide, 96%

Catalog No. p-7030834
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3170703 1 g
AAH3170706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3170703 Supplier Thermo Scientific Chemicals Supplier No. H3170703

CAS: 51770-71-9 | C8H11NO4S | 217.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51770-71-9
Molecular Formula C8H11NO4S
Molecular Weight (g/mol) 217.24
MDL Number MFCD06147001
InChI Key MGHCDRVTMABICG-UHFFFAOYSA-N
Synonym 2,4-dimethoxybenzene-1-sulfonamide, benzenesulfonamide,2,4-dimethoxy
PubChem CID 6469795
IUPAC Name 2,4-dimethoxybenzenesulfonamide
SMILES COC1=CC(OC)=C(C=C1)S(N)(=O)=O

Specifications

Melting Point 166°C to 170°C
Quantity 1 g
Formula Weight 217.25
Percent Purity 96%
Chemical Name or Material 2,4-Dimethoxybenzenesulfonamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only