CAS RN 51770-71-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,4-Dimethoxybenzenesulfonamide, 96%
CAS: 51770-71-9 Molecular Formula: C8H11NO4S Molecular Weight (g/mol): 217.24 MDL Number: MFCD06147001 InChI Key: MGHCDRVTMABICG-UHFFFAOYSA-N Synonym: 2,4-dimethoxybenzene-1-sulfonamide,benzenesulfonamide,2,4-dimethoxy PubChem CID: 6469795 IUPAC Name: 2,4-dimethoxybenzenesulfonamide SMILES: COC1=CC(OC)=C(C=C1)S(N)(=O)=O