missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxyaniline 98.0+%, TCI America™

Catalog No. p-7127048
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D198225G 25 g
D1982100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D198225G Supplier TCI America Supplier No. D198225G
Only null left

Chemical Identifiers

CAS 2735-04-8
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.18
MDL Number MFCD00008371
InChI Key GEQNZVKIDIPGCO-UHFFFAOYSA-N
Synonym benzenamine, 2,4-dimethoxy, aniline, 2,4-dimethoxy, 2,4-dimethoxyphenylamine, unii-i079354gsi, benzenamine, 2,4-dimethoxy-, hydrochloride, 2,4-domethoxyaniline, 2,4-dimethoxy-anilin, 2, 4-dimethoxyaniline, 2,4 dimethoxy aniline, 2,4-dimethoxy aniline
PubChem CID 17652
IUPAC Name 2,4-dimethoxyaniline
SMILES COC1=CC(=C(C=C1)N)OC

Specifications

Quantity 25 g
Formula Weight 153.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material 2,4-Dimethoxyaniline
EINECSNumber (3)-0616
RTECSNumber BX4200000
TSCA Yes