accessibility menu, dialog, popup

UserName

H3CONH2OH3C

CAS RN 2735-04-8

IUPAC Name: 2,4-dimethoxyaniline
Synonyms: 2,4-dimethoxyaniline
Molecular Weight (g/mol): 153.18
Molecular Formula: C8H11NO2
InChi Key: GEQNZVKIDIPGCO-UHFFFAOYSA-N
SMILES: COC1=CC=C(N)C(OC)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results