Learn More
2,4-Dihydroxybenzophenone, 99%
CAS: 131-56-6 | C13H10O3 | 214.22 g/mol
Supplier: Thermo Scientific Chemicals A1374922
Description
Novel mordent and disperse azo dyes were prepared by the coupling of various diazo solutions of aromatic amines with 2, 4-dihydroxybenzophenone. The ultraviolet absorbing monomers were synthesized by reaction of 2, 4-dihydroxybenzophenone with glycidyl acrylate and glycidyl methacrylate. Two new polymerizable stabilizers, 4-benzoyl-2-(α-piperidino-2-chlorobenzyl)-3-hydroxyphenyl acrylate (BPBHA) and corresponding methacrylate (BPBHMA), were synthesized and characterized from 2,4-Dihydroxybenzophenone. a polymerizable UV-stabilizer, 2-hydroxy-4-(3-methacryloxy-2-hydroxylpropoxy) benzophenone (BPMA), was synthesized using 2, 4-dihydroxybenzophenone (UV-0) and glycidyl methacrylate (GMA).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNovel mordent and disperse azo dyes were prepared by the coupling of various diazo solutions of aromatic amines with 2, 4-dihydroxybenzophenone. The ultraviolet absorbing monomers were synthesized by reaction of 2, 4-dihydroxybenzophenone with glycidyl acrylate and glycidyl methacrylate. Two new polymerizable stabilizers, 4-benzoyl-2-(α-piperidino-2-chlorobenzyl)-3-hydroxyphenyl acrylate (BPBHA) and corresponding methacrylate (BPBHMA), were synthesized and characterized from 2,4-Dihydroxybenzophenone. a polymerizable UV-stabilizer, 2-hydroxy-4-(3-methacryloxy-2-hydroxylpropoxy) benzophenone (BPMA), was synthesized using 2, 4-dihydroxybenzophenone (UV-0) and glycidyl methacrylate (GMA).
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
| 2,4-Dihydroxybenzophenone | |
| 144°C to 147°C | |
| 194°C (1 mmHg) | |
| C13H10O3 | |
| 100 g | |
| 14,1106 | |
| Insoluble in water. | |
| C1=CC=C(C=C1)C(=O)C2=C(C=C(C=C2)O)O | |
| 214.22 | |
| CHEBI:34240 | |
| 99% |
| 131-56-6 | |
| 1.32 | |
| 125°C (257°F) | |
| MFCD00002277 | |
| 1311566 | |
| 2,4-dihydroxybenzophenone, 2,4-dihydroxyphenyl phenyl methanone, benzophenone-1, benzoresorcinol, resbenzophenone, inhibitor dhbp, uvinul 400, advastab 48, uvistat 12, methanone, 2,4-dihydroxyphenyl phenyl | |
| ZXDDPOHVAMWLBH-UHFFFAOYSA-N | |
| (2,4-dihydroxyphenyl)-phenylmethanone | |
| 8572 | |
| 214.22 |
Chemical Identifiers
| 131-56-6 | |
| 214.22 | |
| ZXDDPOHVAMWLBH-UHFFFAOYSA-N | |
| 8572 | |
| (2,4-dihydroxyphenyl)-phenylmethanone |
| C13H10O3 | |
| MFCD00002277 | |
| 2,4-dihydroxybenzophenone, 2,4-dihydroxyphenyl phenyl methanone, benzophenone-1, benzoresorcinol, resbenzophenone, inhibitor dhbp, uvinul 400, advastab 48, uvistat 12, methanone, 2,4-dihydroxyphenyl phenyl | |
| CHEBI:34240 | |
| C1=CC=C(C=C1)C(=O)C2=C(C=C(C=C2)O)O |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
H319-H361
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 205-029-4
RTECSNumber : DJ0700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only