missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxybenzophenone, 99%

Catalog No. p-7021425
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1374922 100 g
AAA1374936 500 g
AAA137490E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1374922 Supplier Thermo Scientific Chemicals Supplier No. A1374922

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 131-56-6 | C13H10O3 | 214.22 g/mol

Novel mordent and disperse azo dyes were prepared by the coupling of various diazo solutions of aromatic amines with 2, 4-dihydroxybenzophenone. The ultraviolet absorbing monomers were synthesized by reaction of 2, 4-dihydroxybenzophenone with glycidyl acrylate and glycidyl methacrylate. Two new polymerizable stabilizers, 4-benzoyl-2-(α-piperidino-2-chlorobenzyl)-3-hydroxyphenyl acrylate (BPBHA) and corresponding methacrylate (BPBHMA), were synthesized and characterized from 2,4-Dihydroxybenzophenone. a polymerizable UV-stabilizer, 2-hydroxy-4-(3-methacryloxy-2-hydroxylpropoxy) benzophenone (BPMA), was synthesized using 2, 4-dihydroxybenzophenone (UV-0) and glycidyl methacrylate (GMA).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Novel mordent and disperse azo dyes were prepared by the coupling of various diazo solutions of aromatic amines with 2, 4-dihydroxybenzophenone. The ultraviolet absorbing monomers were synthesized by reaction of 2, 4-dihydroxybenzophenone with glycidyl acrylate and glycidyl methacrylate. Two new polymerizable stabilizers, 4-benzoyl-2-(α-piperidino-2-chlorobenzyl)-3-hydroxyphenyl acrylate (BPBHA) and corresponding methacrylate (BPBHMA), were synthesized and characterized from 2,4-Dihydroxybenzophenone. a polymerizable UV-stabilizer, 2-hydroxy-4-(3-methacryloxy-2-hydroxylpropoxy) benzophenone (BPMA), was synthesized using 2, 4-dihydroxybenzophenone (UV-0) and glycidyl methacrylate (GMA).

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 131-56-6
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00002277
InChI Key ZXDDPOHVAMWLBH-UHFFFAOYSA-N
Synonym 2,4-dihydroxybenzophenone, 2,4-dihydroxyphenyl phenyl methanone, benzophenone-1, benzoresorcinol, resbenzophenone, inhibitor dhbp, uvinul 400, advastab 48, uvistat 12, methanone, 2,4-dihydroxyphenyl phenyl
PubChem CID 8572
ChEBI CHEBI:34240
IUPAC Name (2,4-dihydroxyphenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=C(C=C(C=C2)O)O

Specifications

Melting Point 144°C to 147°C
Density 1.32
Boiling Point 194°C (1 mmHg)
Flash Point 125°C (257°F)
Quantity 100 g
Beilstein 1311566
Merck Index 14,1106
Solubility Information Insoluble in water.
Formula Weight 214.22
Percent Purity 99%
Chemical Name or Material 2,4-Dihydroxybenzophenone
Show More Show Less
Hazard Category H319-H361
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 205-029-4
RTECSNumber DJ0700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only