missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenylglyoxal hydrate, 95%, dry wt. basis

Catalog No. AAL2026603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2026603 1 g
AAL2026606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2026603 Supplier Thermo Scientific Chemicals Supplier No. L2026603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 79784-36-4 | C8H4F2O2 | 170.12 g/mol

Used in the synthesis of biologically active 1,2,4-triazine derivatives. Also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of biologically active 1,2,4-triazine derivatives. Also used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 79784-36-4
Molecular Formula C8H4F2O2
Molecular Weight (g/mol) 170.12
MDL Number MFCD04038287
InChI Key AZBQYELTMQNVST-UHFFFAOYSA-N
Synonym 2,4-difluorophenylglyoxal hydrate, 2-2,4-difluorophenyl-2-oxoacetaldehyde hydrate, 2,4-difluorophenylglyoxal monohydrate, benzeneacetaldehyde,2,4-difluoro-a-oxo, 2-2,4-difluorophenyl-2-oxoethanal, hydrate, 2-2,4-bis fluoranyl phenyl-2-oxidanylidene-ethanal hydrate
PubChem CID 2782300
IUPAC Name 2-(2,4-difluorophenyl)-2-oxoacetaldehyde;hydrate
SMILES FC1=CC(F)=C(C=C1)C(=O)C=O

Specifications

Melting Point 51°C to 54°C
Quantity 1 g
Beilstein 4387465
Solubility Information Soluble in water.
Formula Weight 170.12 (Anhydrous)
Percent Purity 95%
Assay (dry wt.)
Chemical Name or Material 2,4-Difluorophenylglyoxal hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only