missing translation for 'onlineSavingsMsg'
Learn More

(2,4-Difluorophenyl)thiourea 98.0+%, TCI America™

Catalog No. D48611G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48611G 1 g
D48615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48611G Supplier TCI America Supplier No. D48611G

Chemical Identifiers

CAS 175277-76-6
Molecular Formula C7H6F2N2S
Molecular Weight (g/mol) 188.196
MDL Number MFCD00041150
InChI Key DZZSKQFBAGZNSH-UHFFFAOYSA-N
Synonym 2,4-difluorophenyl thiourea, 1-2,4-difluorophenyl thiourea, 1-2,4-difluorophenyl-2-thiourea, n-2,4-difluorophenyl thiourea, thiourea,n-2,4-difluorophenyl, amino 2,4-difluorophenyl amino methane-1-thione, acmc-1bxvl, buttpark 12118-31, difluorophenylthioacetonitrile, 2,4-difluoro-phenyl-thiourea
PubChem CID 2734207
IUPAC Name (2,4-difluorophenyl)thiourea
SMILES C1=CC(=C(C=C1F)F)NC(=S)N

Specifications

Melting Point 168°C
Quantity 1 g
UN Number 2811
Formula Weight 188.20
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (2,4-Difluorophenyl)thiourea