CAS RN 175277-76-6
IUPAC Name:
(2,4-difluorophenyl)thiourea
Synonyms:
2,4-difluorophenylthiourea
Molecular Weight (g/mol):
188.2
Molecular Formula:
C7H6F2N2S
InChi Key:
DZZSKQFBAGZNSH-UHFFFAOYSA-N
SMILES:
NC(=S)NC1=CC=C(F)C=C1F
1
–
1
of
1
results
1
(2,4-Difluorophenyl)thiourea 98.0+%, TCI America™
CAS: 175277-76-6 Molecular Formula: C7H6F2N2S Molecular Weight (g/mol): 188.196 MDL Number: MFCD00041150 InChI Key: DZZSKQFBAGZNSH-UHFFFAOYSA-N Synonym: 2,4-difluorophenyl thiourea,1-2,4-difluorophenyl thiourea,1-2,4-difluorophenyl-2-thiourea,n-2,4-difluorophenyl thiourea,thiourea,n-2,4-difluorophenyl,amino 2,4-difluorophenyl amino methane-1-thione,acmc-1bxvl,buttpark 12118-31,difluorophenylthioacetonitrile,2,4-difluoro-phenyl-thiourea PubChem CID: 2734207 IUPAC Name: (2,4-difluorophenyl)thiourea SMILES: C1=CC(=C(C=C1F)F)NC(=S)N