missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenyl Isothiocyanate 98.0+%, TCI America™

Catalog No. D34541G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D34541G Supplier TCI America Supplier No. D34541G

Chemical Identifiers

CAS 141106-52-7
Molecular Formula C7H3F2NS
Molecular Weight (g/mol) 171.165
MDL Number MFCD00041044
InChI Key ABGGPKIFVAIRGU-UHFFFAOYSA-N
Synonym 2,4-difluorophenyl isothiocyanate, 2,4-difluoro isothiocyanato benzene, 2,4-difluoro-1-isothiocyanato-benzene, 2,4-difluorophenylisothiocyanate, isothiocyanic acid 2,4-difluorophenyl ester, benzene, 2,4-difluoro-1-isothiocyanato, benzene, 2,4-difluoro-1-isothiocyanato-9ci, 2,4-difluorobenzenisothiocyanate, pubchem4451, acmc-209clq
PubChem CID 518879
IUPAC Name 2,4-difluoro-1-isothiocyanatobenzene
SMILES C1=CC(=C(C=C1F)F)N=C=S

Specifications

Color Yellow
Boiling Point 86°C
Quantity 1 g
UN Number 2810
Formula Weight 171.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Difluorophenyl Isothiocyanate