accessibility menu, dialog, popup

UserName

FNCSF

CAS RN 141106-52-7

IUPAC Name: 2,4-difluoro-1-isothiocyanatobenzene
Synonyms: 2,4-difluoro-1-isothiocyanatobenzene
Molecular Weight (g/mol): 171.16
Molecular Formula: C7H3F2NS
InChi Key: ABGGPKIFVAIRGU-UHFFFAOYSA-N
SMILES: FC1=CC=C(N=C=S)C(F)=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)